Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other R3HCC1L SIRNA Assays ›
Gene Symbol:
R3HCC1L
Gene Name:
R3H domain and coiled-coil containing 1 like
Gene Aliases:
C10orf28, GIDRP86, GIDRP88, PSORT
Chromosome Location:
Chr.10: 98134608 - 98244897 on Build GRCh38
UniGene ID:
Hs.419800
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS120584
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: R3HCC1L
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256619.1
NP_001243548.1
5
1539
NM_001256620.1
NP_001243549.1
4
1488
NM_014472.4
NP_055287.4
4
1484
NM_138469.2
NP_612478.2
5
1581
XM_011539639.1
XP_011537941.1
5
1796
XM_011539640.1
XP_011537942.1
6
1601
XM_011539641.1
XP_011537943.1
5
1553
XM_011539642.1
XP_011537944.1
6
1893
XM_011539643.1
XP_011537945.1
4
1480
XM_011539644.1
XP_011537946.1
4
1484
XM_011539645.2
XP_011537947.1
4
1727
XM_011539646.1
XP_011537948.1
3
1359
XM_011539647.1
XP_011537949.1
3
1361
XM_011539648.1
XP_011537950.1
2
1234
XM_011539649.1
XP_011537951.1
5
1796
XM_017016074.1
XP_016871563.1
4
2692
XM_017016075.1
XP_016871564.1
2
1234
XM_017016076.1
XP_016871565.1
5
1528
XM_017016077.1
XP_016871566.1
7
1691
XM_017016078.1
XP_016871567.1
6
1613
XM_017016079.1
XP_016871568.1
6
1627
XM_017016080.1
XP_016871569.1
4
1533
XM_017016081.1
XP_016871570.1
5
1615
XM_017016082.1
XP_016871571.1
3
1407
XM_017016083.1
XP_016871572.1
4
1423
GenBank mRNA
AF525304.1
-
5
1492
AK292546.1
-
4
1483
BC098268.1
-
5
1522
40026913
BC098359.1
-
4
1425
40026912
BC099848.1
-
4
1425
40026916
Back To Top
More Information
Gene Ontology Categories:
Process(es)
nucleotide binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0