Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SGSM3 SIRNA Assays ›
          Gene Symbol:
          SGSM3
          Gene Name:
          small G protein signaling modulator 3
          Gene Aliases:
          MAP, RABGAP5, RUSC3, RUTBC3, RabGAP-5, rabGAPLP
          Chromosome Location:
          Chr.22: 40370530 - 40410289 on Build GRCh38
          UniGene ID:
          Hs.474914
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS120681
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SGSM3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001301849.1 NP_001288778.1 16 1826
          NM_015705.5 NP_056520.2 17 2049
          XM_005261572.2 XP_005261629.1 17 2076
          XM_005261573.3 XP_005261630.1 17 2081
          XM_005261574.4 XP_005261631.1 17 2081
          XM_005261575.1 XP_005261632.1 16 1944
          XM_011530158.1 XP_011528460.1 16 1900
          XM_011530159.1 XP_011528461.1 17 1964
          XM_011530160.1 XP_011528462.1 16 1881
          XM_011530161.1 XP_011528463.1 17 2073
          XM_011530162.2 XP_011528464.1 17 1959
          XM_017028775.1 XP_016884264.1 16 1868
          XM_017028776.1 XP_016884265.1 16 1937
          XM_017028777.1 XP_016884266.1 16 1937
          XM_017028778.1 XP_016884267.1 15 1800
          XM_017028779.1 XP_016884268.1 16 1990
          XM_017028780.1 XP_016884269.1 16 1979
          GenBank mRNA AB196956.1 - 15 1818
          AB275763.1 - 16 1922
          AB449880.1 - 15 1811
          AK000006.1 - 7 2959
          AK295469.1 - 17 1924
          AK297599.1 - 16 1897
          AK301042.1 - 16 1739
          AY308849.1 - 17 1984
          BC008078.2 - 17 1975 3027777
          CR456441.1 - 16 1847

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          GTPase-activating protein

          Gene Ontology Categories:

          Function(s) Process(es)

          intracellular protein transport
          cell cycle arrest
          regulation of vesicle fusion
          regulation of Rab protein signal transduction
          Rap protein signal transduction
          plasma membrane to endosome transport
          activation of GTPase activity
          regulation of cilium assembly
          GTPase activator activity
          protein binding
          Rab GTPase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-km2td:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0