Search
Search

ZBTB44| Gene Symbol: ZBTB44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301098.1 | NP_001288027.1 | 2 | 793 | ||
| NM_001301099.1 | NP_001288028.1 | 2 | 793 | |||
| NM_014155.4 | NP_054874.3 | 2 | 793 | |||
| XM_005271521.4 | XP_005271578.1 | 2 | 989 | |||
| XM_005271522.4 | XP_005271579.1 | 2 | 868 | |||
| XM_006718825.3 | XP_006718888.1 | 2 | 868 | |||
| XM_006718826.3 | XP_006718889.1 | 2 | 865 | |||
| XM_011542789.2 | XP_011541091.1 | 2 | 866 | |||
| XM_017017623.1 | XP_016873112.1 | 2 | 865 | |||
| XM_017017624.1 | XP_016873113.1 | 2 | 865 | |||
| GenBank mRNA | AF161548.1 | - | 2 | 512 | ||
| AK314701.1 | - | 2 | 670 | |||
| BC030580.1 | - | 2 | 677 | 4823171 | ||
| BC049375.1 | - | 2 | 698 | 5271784 | ||
| BC050723.1 | - | 1 | 415 | 4445874 | ||
| BC071729.1 | - | 2 | 772 | 5163748 | ||