Search
Search

TFCP2L1| Gene Symbol: TFCP2L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014553.2 | NP_055368.1 | 3, 4 | 390 | ||
| XM_017003902.1 | XP_016859391.1 | 3, 4 | 398 | |||
| XM_017003904.1 | XP_016859393.1 | 3, 4 | 398 | |||
| XM_017003905.1 | XP_016859394.1 | 3, 4 | 398 | |||
| GenBank mRNA | AF198488.1 | - | 3, 4 | 387 | ||
| AL137740.1 | - | 3, 4 | 391 | |||
| BC064698.1 | - | 3, 4 | 390 | 30347863 | ||