Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: NME7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013330.4 | NP_037462.1 | 6 | 713 | ||
NM_197972.2 | NP_932076.1 | 6 | 682 | |||
NR_104229.1 | 6 | 713 | ||||
XM_005245106.4 | XP_005245163.1 | 4 | 291 | |||
XM_017001099.1 | XP_016856588.1 | 6 | 699 | |||
XM_017001100.1 | XP_016856589.1 | 4 | 279 | |||
GenBank mRNA | AB209049.1 | - | 6 | 522 | ||
AF153191.2 | - | 6 | 552 | |||
AK094513.1 | - | 6 | 665 | |||
AK290701.1 | - | 6 | 547 | |||
AK301917.1 | - | 6 | 696 | |||
BC006983.1 | - | 6 | 544 | 3997152 |