Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PADI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013358.2 | NP_037490.2 | 2 | 313 | ||
XM_011541307.1 | XP_011539609.1 | 2 | 316 | |||
XM_017001101.1 | XP_016856590.1 | 2 | 216 | |||
XM_017001102.1 | XP_016856591.1 | 2 | 216 | |||
XM_017001103.1 | XP_016856592.1 | 2 | 316 | |||
GenBank mRNA | AB033768.2 | - | 2 | 304 | ||
AK026652.1 | - | 2 | 315 | |||
AK225541.1 | - | 2 | 315 | |||
AK314644.1 | - | 2 | 314 | |||
BC130574.1 | - | 2 | 292 | 40146605 | ||
BC136402.1 | - | 2 | 292 | 9020389 |