Search
Search

LRP12| Gene Symbol: LRP12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135703.2 | NP_001129175.1 | 4 | 1432 | ||
| NM_013437.4 | NP_038465.1 | 5 | 1489 | |||
| GenBank mRNA | AB208957.1 | - | 3 | 1164 | ||
| AF166350.1 | - | 5 | 1404 | |||
| AK289752.1 | - | 5 | 1393 | |||
| AK299374.1 | - | 4 | 1429 | |||
| BC032109.2 | - | 5 | 1365 | 4842797 | ||