Search Thermo Fisher Scientific
Gene Symbol: SLC2A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271712.1 | NP_001258641.1 | 7 | 994 | ||
NM_014580.4 | NP_055395.2 | 9 | 1364 | |||
NR_073416.1 | 6 | 797 | ||||
XM_006717084.3 | XP_006717147.1 | 9 | 1359 | |||
XM_011518602.2 | XP_011516904.1 | 9 | 1280 | |||
XM_011518603.2 | XP_011516905.1 | 7 | 1019 | |||
XM_011518604.2 | XP_011516906.1 | 7 | 943 | |||
GenBank mRNA | AJ245937.1 | - | 9 | 1274 | ||
BC019043.2 | - | 9 | 1316 | 4641145 | ||
BM044230.1 | - | 6 | 698 | |||
BX395379.2 | - | 7 | 918 | |||
Y17801.1 | - | 9 | 1297 |