Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VILL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015873.3 | NP_056957.3 | 17 | 2272 | ||
XM_005265191.3 | XP_005265248.1 | 18 | 2380 | |||
XM_005265192.4 | XP_005265249.1 | 18 | 3515 | |||
XM_006713184.3 | XP_006713247.1 | 19 | 2380 | |||
XM_011533770.2 | XP_011532072.1 | 18 | 2536 | |||
GenBank mRNA | AB209574.1 | - | 3 | 3729 | ||
BC000243.1 | - | 14 | 1808 | 3356294 | ||
BC004300.1 | - | 18 | 2302 | 3622951 | ||
D88154.1 | - | 13 | 1678 | |||
KU178619.1 | - | 13 | 1676 | |||
KU178620.1 | - | 12 | 1530 |