Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ERGIC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016570.2 | NP_057654.2 | 5 | 447 | ||
GenBank mRNA | AF183410.1 | - | 5 | 398 | ||
AF216751.1 | - | 5 | 402 | |||
AF302767.1 | - | 5 | 400 | |||
AK074520.1 | - | 5 | 422 | |||
AK222799.1 | - | 5 | 387 | |||
AL834128.2 | - | 5 | 388 | |||
BC000887.1 | - | 5 | 400 | 3462408 | ||
BC107794.1 | - | 5 | 404 | 30319807 | ||
BF681475.1 | - | 475 | ||||
BI547753.1 | - | 5 | 451 |