Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other UCHL5 SIRNA Assays ›
          Gene Symbol:
          UCHL5
          Gene Name:
          ubiquitin C-terminal hydrolase L5
          Gene Aliases:
          CGI-70, INO80R, UCH-L5, UCH37
          Chromosome Location:
          Chr.1: 193012366 - 193060140 on Build GRCh38
          UniGene ID:
          Hs.145469
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS122114
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price 459.00
          Your Price
          Online offer:
          459.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: UCHL5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001199261.1 NP_001186190.1 3, 4 386
          NM_001199262.1 NP_001186191.1 3, 4 386
          NM_001199263.1 NP_001186192.1 3, 4 386
          NM_015984.3 NP_057068.1 3, 4 386
          NR_037607.1 3, 4 652
          XM_005245244.4 XP_005245301.1 3, 4 469
          XM_005245246.4 XP_005245303.1 3, 4 469
          XM_005245247.4 XP_005245304.1 3, 4 321
          XM_005245248.4 XP_005245305.1 3, 4 344
          XM_005245249.4 XP_005245306.1 3, 4 240
          XM_006711366.3 XP_006711429.1 3, 4 469
          XM_006711367.3 XP_006711430.1 3, 4 469
          XM_006711368.3 XP_006711431.1 3, 4 469
          XM_006711369.2 XP_006711432.1 3, 4 469
          XM_006711370.3 XP_006711433.3 3, 4 422
          XM_006711371.3 XP_006711434.1 3, 4 240
          XM_011509604.1 XP_011507906.1 3, 4 469
          XM_011509607.2 XP_011507909.1 3, 4 280
          XM_011509608.2 XP_011507910.1 4, 5 641
          XM_011509609.2 XP_011507911.1 4, 5 545
          XM_017001429.1 XP_016856918.1 3, 4 469
          XM_017001430.1 XP_016856919.1 3, 4 469
          XM_017001431.1 XP_016856920.1 3, 4 469
          XM_017001432.1 XP_016856921.1 3, 4 469
          XM_017001433.1 XP_016856922.1 3, 4 469
          XM_017001434.1 XP_016856923.1 3, 4 422
          XM_017001435.1 XP_016856924.1 3, 4 422
          XM_017001436.1 XP_016856925.1 4, 5 641
          XM_017001437.1 XP_016856926.1 4, 5 641
          XM_017001438.1 XP_016856927.1 3, 4 264
          XM_017001439.1 XP_016856928.1 3, 4 685
          XM_017001440.1 XP_016856929.1 3, 4 649
          XM_017001441.1 XP_016856930.1 3, 4 625
          XM_017001442.1 XP_016856931.1 3, 4 240
          XM_017001443.1 XP_016856932.1 3, 4 240
          XM_017001444.1 XP_016856933.1 3, 4 634
          GenBank mRNA AF147717.1 - 3, 4 384
          AF151828.1 - 3, 4 266
          AF157320.1 - 3, 4 401
          AK024751.1 - 3, 4 332
          AK225794.1 - 3, 4 319
          AK301381.1 - 3, 4 652
          AK314445.1 - 3, 4 330
          BC015521.1 - 3, 4 319 3912584
          BC025369.2 - 3, 4 364 3937407
          BT006790.1 - 3, 4 253
          DA255130.1 - 3, 4 386

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          cysteine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA repair
          DNA recombination
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          ubiquitin-dependent protein catabolic process
          negative regulation of endopeptidase activity
          protein deubiquitination
          lateral ventricle development
          midbrain development
          forebrain morphogenesis
          regulation of proteasomal protein catabolic process
          thiol-dependent ubiquitin-specific protease activity
          endopeptidase inhibitor activity
          protein binding
          poly(A) RNA binding
          proteasome binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0