Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SUFU SIRNA Assays ›
          Gene Symbol:
          SUFU
          Gene Name:
          SUFU negative regulator of hedgehog signaling
          Gene Aliases:
          PRO1280, SUFUH, SUFUXL
          Chromosome Location:
          Chr.10: 102502879 - 102633458 on Build GRCh38
          UniGene ID:
          Hs.404089
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS122523
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SUFU
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001178133.1 NP_001171604.1 2 400
          NM_016169.3 NP_057253.2 2 400
          XM_011539858.2 XP_011538160.1 2 400
          XM_011539860.2 XP_011538162.1 2 400
          XM_011539861.2 XP_011538163.1 2 400
          XM_011539863.2 XP_011538165.1 2 135
          XM_011539864.2 XP_011538166.1 2 400
          XM_017016323.1 XP_016871812.1 2 630
          GenBank mRNA AF144231.1 - 2 355
          AF159447.1 - 2 355
          AF172319.1 - 2 205
          AF175770.1 - 2 375
          AF222345.1 - 2 283
          AK312319.1 - 2 390
          AY358550.1 - 2 355
          BC013291.2 - 2 376 3533158
          DA248208.1 - 2 400

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_001025391, NM_015752, AK047603, AJ308626, AJ131692, and others.

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          skeletal system development
          neural tube closure
          heart looping
          ventricular septum development
          regulation of transcription, DNA-templated
          proteolysis
          signal transduction
          multicellular organism development
          smoothened signaling pathway involved in ventral spinal cord interneuron specification
          smoothened signaling pathway involved in spinal cord motor neuron cell fate specification
          aorta development
          negative regulation of transcription factor import into nucleus
          cytoplasmic sequestering of transcription factor
          negative regulation of sequence-specific DNA binding transcription factor activity
          skin development
          negative regulation of osteoblast differentiation
          negative regulation of smoothened signaling pathway
          coronary vasculature development
          negative regulation of smoothened signaling pathway involved in dorsal/ventral neural tube patterning
          negative regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          transcription corepressor activity
          signal transducer activity
          protein binding
          beta-catenin binding
          transcription factor binding
          protein kinase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0