Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MIS18A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018944.2 | NP_061817.1 | 5 | 685 | ||
GenBank mRNA | AF231921.1 | - | 5 | 657 | ||
AF387845.1 | - | 5 | 655 | |||
AK075281.1 | - | 5 | 657 | |||
AK312073.1 | - | 5 | 776 | |||
AW962893.1 | - | 3 | 139 | |||
BC042917.1 | - | 5 | 661 | 5934336 | ||
BU567844.1 | - | 3 | 252 |