Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SH3TC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318480.1 | NP_001305409.1 | 18 | 4166 | ||
NM_018986.4 | NP_061859.3 | 18 | 4083 | |||
NR_134639.1 | 15 | 3646 | ||||
XM_006713889.2 | XP_006713952.1 | 15 | 3501 | |||
XM_006713891.3 | XP_006713954.1 | 12 | 3143 | |||
XM_011513488.1 | XP_011511790.1 | 18 | 4060 | |||
XM_011513489.2 | XP_011511791.1 | 16 | 3683 | |||
XM_017008300.1 | XP_016863789.1 | 18 | 4192 | |||
XM_017008301.1 | XP_016863790.1 | 18 | 3875 | |||
XM_017008302.1 | XP_016863791.1 | 15 | 3585 | |||
GenBank mRNA | AK000363.1 | - | 14 | 3470 | ||
AK074402.1 | - | 18 | 3960 | |||
AK093562.1 | - | 7 | 2004 | |||
AK126165.1 | - | 18 | 4039 | |||
AK128258.1 | - | 4 | 3544 | |||
AK222865.1 | - | 14 | 3471 |