Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BNC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317939.1 | NP_001304868.1 | 7 | 3127 | ||
NM_001317940.1 | NP_001304869.1 | 6 | 2977 | |||
NM_017637.5 | NP_060107.3 | 7 | 3197 | |||
XM_011517924.1 | XP_011516226.1 | 8 | 3262 | |||
XM_017014816.1 | XP_016870305.1 | 11 | 5648 | |||
XM_017014817.1 | XP_016870306.1 | 9 | 3722 | |||
XM_017014818.1 | XP_016870307.1 | 9 | 3712 | |||
XM_017014819.1 | XP_016870308.1 | 8 | 3655 | |||
XM_017014820.1 | XP_016870309.1 | 8 | 3649 | |||
XM_017014821.1 | XP_016870310.1 | 7 | 3569 | |||
XM_017014822.1 | XP_016870311.1 | 7 | 3258 | |||
XM_017014823.1 | XP_016870312.1 | 7 | 8027 | |||
XM_017014824.1 | XP_016870313.1 | 6 | 3579 | |||
XM_017014825.1 | XP_016870314.1 | 10 | 3754 | |||
XM_017014829.1 | XP_016870318.1 | 7 | 3659 | |||
GenBank mRNA | AK000050.1 | - | 2 | 981 | ||
AK025882.1 | - | 2 | 726 | |||
AK092247.1 | - | 4 | 2661 | |||
AK131398.1 | - | 7 | 3079 | |||
AK299073.1 | - | 6 | 2947 | |||
AY438376.1 | - | 5 | 2842 | |||
BX641030.1 | - | 2 | 2227 | |||
CR933649.1 | - | 7 | 3084 |