Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other AHI1 SIRNA Assays ›
          Gene Symbol:
          AHI1
          Gene Name:
          Abelson helper integration site 1
          Gene Aliases:
          AHI-1, JBTS3, ORF1, dJ71N10.1
          Chromosome Location:
          Chr.6: 135283972 - 135497775 on Build GRCh38
          UniGene ID:
          Hs.386684
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS123281
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: AHI1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001134830.1 NP_001128302.1 21, 22 3354
          NM_001134831.1 NP_001128303.1 23, 24 3501
          NM_017651.4 NP_060121.3 22, 23 3416
          XM_011535910.2 XP_011534212.1 24, 25 3645
          XM_011535911.2 XP_011534213.1 22, 23 3375
          XM_017010977.1 XP_016866466.1 23, 24 3620
          XM_017010978.1 XP_016866467.1 22, 23 3318
          XM_017010979.1 XP_016866468.1 22, 23 3318
          XM_017010980.1 XP_016866469.1 22, 23 3318
          XM_017010981.1 XP_016866470.1 22, 23 3453
          XM_017010982.1 XP_016866471.1 22, 23 3318
          XM_017010983.1 XP_016866472.1 22, 23 3318
          GenBank mRNA AJ459824.1 - 22, 23 3396
          AJ606362.1 - 21, 22 3334
          AK024085.1 - 10, 11 1309
          AK092262.1 - 21, 22 3327
          AL136797.1 - 20, 21 3135
          BC015019.1 - 12, 13 1615
          BC094800.1 - 23, 24 3424 4823262
          DQ090887.1 - 23, 24 3455

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          morphogenesis of a polarized epithelium
          heart looping
          positive regulation of receptor internalization
          transmembrane receptor protein tyrosine kinase signaling pathway
          central nervous system development
          retina layer formation
          vesicle-mediated transport
          positive regulation of polarized epithelial cell differentiation
          hindbrain development
          cellular protein localization
          cloaca development
          photoreceptor cell outer segment organization
          pronephric nephron tubule morphogenesis
          pronephric duct morphogenesis
          cilium assembly
          negative regulation of apoptotic process
          positive regulation of transcription from RNA polymerase II promoter
          regulation of behavior
          cilium morphogenesis
          specification of axis polarity
          Kupffer's vesicle development
          left/right axis specification
          otic vesicle development
          protein binding
          identical protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0