Search Thermo Fisher Scientific
Gene Symbol: TXNL4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142317.1 | NP_001135789.1 | 2 | 281 | ||
NM_001142318.1 | NP_001135790.1 | 2 | 179 | |||
NM_001324354.1 | NP_001311283.1 | 2 | 341 | |||
NM_001324355.1 | NP_001311284.1 | 2 | 341 | |||
NM_017853.2 | NP_060323.1 | 2 | 330 | |||
XM_017023377.1 | XP_016878866.1 | 2 | 346 | |||
GenBank mRNA | AK000518.1 | - | 2 | 305 | ||
AK222889.1 | - | 2 | 297 | |||
AK308891.1 | - | 2 | 247 | |||
AY566808.1 | - | 1 | 19 | |||
BC009646.1 | - | 2 | 271 | 3458545 | ||
BM849401.1 | - | 2 | 242 | |||
BP355646.1 | - | 2 | 330 | |||
CD517462.1 | - | 2 | 227 | |||
CR457240.1 | - | 1 | 19 | |||
DA499499.1 | - | 2 | 281 |