Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: THG1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317824.1 | NP_001304753.1 | 6 | 831 | ||
NM_001317825.1 | NP_001304754.1 | 6 | 922 | |||
NM_001317826.1 | NP_001304755.1 | 7 | 894 | |||
NM_017872.4 | NP_060342.2 | 6 | 926 | |||
XM_017009603.1 | XP_016865092.1 | 6 | 861 | |||
GenBank mRNA | AF086486.1 | - | 1 | 102 | ||
AK000553.1 | - | 6 | 902 | |||
AK021663.1 | - | 6 | 877 | |||
AK023355.1 | - | 5 | 540 | |||
AK223119.1 | - | 6 | 895 | |||
AK304590.1 | - | 6 | 802 | |||
AL136669.1 | - | 6 | 894 | |||
AY463216.1 | - | 6 | 1000 | |||
BC001523.2 | - | 6 | 872 | 3010614 | ||
BC001852.2 | - | 6 | 872 | 3010614 | ||
BC023521.2 | - | 6 | 892 | 4126840 | ||
CR533503.1 | - | 6 | 874 |