Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PHIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017934.5 | NP_060404.3 | 18 | 2223 | ||
GenBank mRNA | AJ303102.1 | - | 18 | 2010 | ||
AK057039.1 | - | 18 | 2165 | |||
AK127816.1 | - | 6 | 857 | |||
BC021905.1 | - | 18 | 2112 | |||
BC137488.1 | - | 18 | 2103 | 9021488 | ||
DQ924532.1 | - | 18 | 1997 |