Search
Search

ARGLU1| Gene Symbol: ARGLU1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018011.3 | NP_060481.3 | 2 | 648 | ||
| GenBank mRNA | AK001016.1 | - | 2 | 646 | ||
| AK303575.1 | - | 3 | 421 | |||
| AK304751.1 | - | 3 | 478 | |||
| AK309941.1 | - | 2 | 267 | |||
| BC050434.1 | - | 2 | 595 | 6016552 | ||
| BC071587.1 | - | 2 | 619 | 5271043 | ||