Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ATG2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006720187.2 | XP_006720250.1 | 33 | 5291 | ||
XM_017021397.1 | XP_016876886.1 | 33 | 5294 | |||
GenBank mRNA | AK001104.1 | - | 5 | 692 | ||
AK128275.1 | - | 14 | 1739 | |||
AK302641.1 | - | 6 | 915 | |||
BC015016.2 | - | 7 | 1022 |