Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006720187.2 | XP_006720250.1 | 33, 34 | 5402 | ||
XM_017021397.1 | XP_016876886.1 | 33, 34 | 5405 | |||
GenBank mRNA | AK001104.1 | - | 5, 6 | 803 | ||
AK128275.1 | - | 14, 15 | 1850 | |||
AK302641.1 | - | 6, 7 | 1026 | |||
BC015016.2 | - | 7, 8 | 1133 |