Search
Search

MAP1S| Gene Symbol: MAP1S | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308363.1 | NP_001295292.1 | 5 | 979 | ||
| NM_018174.5 | NP_060644.4 | 5 | 873 | |||
| XM_017026930.1 | XP_016882419.1 | 5 | 973 | |||
| XM_017026931.1 | XP_016882420.1 | 5 | 1214 | |||
| GenBank mRNA | AJ440784.1 | - | 5 | 825 | ||
| AK023118.1 | - | 1 | 16 | |||
| AK027623.1 | - | 5 | 825 | |||
| AK292555.1 | - | 5 | 843 | |||
| AK294936.1 | - | 5 | 850 | |||
| AK308975.1 | - | 5 | 773 | |||
| AL834233.1 | - | 5 | 828 | |||
| BC006358.2 | - | 2 | 533 | |||
| BC008806.2 | - | 1 | 65 | |||
| BC067115.1 | - | 5 | 828 | |||
| BC080547.1 | - | 5 | 827 | 5925583 | ||
| BC113952.1 | - | 5 | 825 | 40027183 | ||
| DQ387861.1 | - | 5 | 831 | |||