Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FGGY SIRNA Assays ›
          Gene Symbol:
          FGGY
          Gene Name:
          FGGY carbohydrate kinase domain containing
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 59296405 - 59762730 on Build GRCh38
          UniGene ID:
          Hs.444301
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS124274
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FGGY
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001113411.1 NP_001106882.1 4, 5 665
          NM_018291.3 NP_060761.3 4, 5 665
          NR_103473.1 5, 6 623
          XM_011541730.1 XP_011540032.1 4, 5 589
          XM_011541731.1 XP_011540033.1 4, 5 523
          XM_011541732.1 XP_011540034.1 4, 5 566
          XM_011541733.1 XP_011540035.1 4, 5 484
          XM_011541735.1 XP_011540037.1 4, 5 484
          XM_017001643.1 XP_016857132.1 4, 5 695
          XM_017001644.1 XP_016857133.1 4, 5 488
          XM_017001645.1 XP_016857134.1 4, 5 589
          XM_017001646.1 XP_016857135.1 4, 5 566
          XM_017001647.1 XP_016857136.1 4, 5 694
          XM_017001648.1 XP_016857137.1 5, 6 508
          XM_017001649.1 XP_016857138.1 4, 5 634
          XM_017001650.1 XP_016857139.1 6, 7 857
          XM_017001651.1 XP_016857140.1 6, 7 713
          XM_017001652.1 XP_016857141.1 4, 5 634
          XM_017001653.1 XP_016857142.1 4, 5 566
          XM_017001656.1 XP_016857145.1 5, 6 701
          XM_017001657.1 XP_016857146.1 4, 5 489
          XM_017001658.1 XP_016857147.1 5, 6 632
          XM_017001659.1 XP_016857148.1 3, 4 522
          XM_017001660.1 XP_016857149.1 3, 4 425
          XM_017001661.1 XP_016857150.1 3, 4 582
          XM_017001662.1 XP_016857151.1 3, 4 267
          XM_017001663.1 XP_016857152.1 3, 4 442
          XM_017001664.1 XP_016857153.1 2, 3 319
          XM_017001665.1 XP_016857154.1 2, 3 226
          XM_017001672.1 XP_016857161.1 3, 4 522
          XM_017001673.1 XP_016857162.1 3, 4 579
          XM_017001675.1 XP_016857164.1 3, 4 522
          XM_017001676.1 XP_016857165.1 3, 4 582
          XM_017001679.1 XP_016857168.1 4, 5 634
          GenBank mRNA AK022237.1 - 3, 4 522
          AK090568.1 - 5, 6 623
          AK315649.1 - 4, 5 550
          BC014947.1 - 4, 5 550 4871664
          BX647626.1 - 4, 5 637
          DA072156.1 - 568
          DA969344.1 - 3, 4 450

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          carbohydrate kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          carbohydrate metabolic process
          phosphorylation
          neuron cellular homeostasis
          molecular_function
          kinase activity
          phosphotransferase activity, alcohol group as acceptor

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline