Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other COA1 SIRNA Assays ›
Gene Symbol:
COA1
Gene Name:
cytochrome c oxidase assembly factor 1 homolog
Gene Aliases:
C7orf44, MITRAC15
Chromosome Location:
Chr.7: 43608452 - 43729541 on Build GRCh38
UniGene ID:
Hs.268887
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS124934
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: COA1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001321197.1
NP_001308126.1
5
620
NM_001321198.1
NP_001308127.1
5
501
NM_001321199.1
NP_001308128.1
5
465
NM_001321200.1
NP_001308129.1
5
521
NM_001321201.1
NP_001308130.1
4
393
NM_018224.3
NP_060694.2
4
400
NR_047701.1
5
465
NR_135580.1
5
501
NR_135581.1
5
501
NR_135582.1
4
400
NR_135583.1
4
400
XM_005249807.4
XP_005249864.1
4
464
XM_006715752.2
XP_006715815.1
5
432
XM_017012413.1
XP_016867902.1
4
391
XM_017012414.1
XP_016867903.1
4
391
XM_017012415.1
XP_016867904.1
5
493
XM_017012416.1
XP_016867905.1
4
391
XM_017012417.1
XP_016867906.1
4
391
XM_017012418.1
XP_016867907.1
4
391
XM_017012419.1
XP_016867908.1
4
391
XM_017012420.1
XP_016867909.1
5
493
XM_017012421.1
XP_016867910.1
5
615
XM_017012422.1
XP_016867911.1
5
504
XM_017012423.1
XP_016867912.1
5
424
XM_017012424.1
XP_016867913.1
4
407
XM_017012425.1
XP_016867914.1
5
508
XM_017012426.1
XP_016867915.1
5
627
XM_017012427.1
XP_016867916.1
6
582
XM_017012428.1
XP_016867917.1
4
407
XM_017012429.1
XP_016867918.1
3
301
XM_017012430.1
XP_016867919.1
5
493
XM_017012431.1
XP_016867920.1
4
407
XM_017012432.1
XP_016867921.1
4
407
GenBank mRNA
AK001665.1
-
5
434
AK021905.1
-
5
570
AK023007.1
-
5
417
AK023053.1
-
5
433
AK097203.1
-
3
1931
AK293043.1
-
5
443
BC056884.1
-
5
392
6159045
BG028456.1
-
422
BG334368.1
-
5
410
BG548154.1
-
5
457
BM918741.1
-
4
373
BQ231567.1
-
4
338
DA742258.1
-
4
382
Back To Top
More Information
Panther Classification:
Molecular Function -
chaperone
Gene Ontology Categories:
Function(s)
Process(es)
translation
mitochondrial respiratory chain complex I assembly
mitochondrial respiratory chain complex IV assembly
RNA binding
structural constituent of ribosome
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0