Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PCID2 SIRNA Assays ›
Gene Symbol:
PCID2
Gene Name:
PCI domain containing 2
Gene Aliases:
F10
Chromosome Location:
Chr.13: 113165002 - 113209466 on Build GRCh38
UniGene ID:
Hs.508769
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS125056
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PCID2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001127202.3
NP_001120674.1
14
1414
NM_001127203.3
NP_001120675.1
14
1414
NM_001258213.2
NP_001245142.1
14
1717
NM_001320655.1
NP_001307584.1
14
1717
NM_001320656.1
NP_001307585.1
14
1576
NM_001320657.1
NP_001307586.1
14
1418
NM_001320659.1
NP_001307588.1
14
1418
NM_001320660.1
NP_001307589.1
14
1721
NM_018386.4
NP_060856.2
14
1414
XM_017020664.1
XP_016876153.1
14
1165
XM_017020665.1
XP_016876154.1
14
1723
XM_017020666.1
XP_016876155.1
14
1218
XM_017020667.1
XP_016876156.1
13
1856
XM_017020668.1
XP_016876157.1
13
1061
XM_017020669.1
XP_016876158.1
13
1902
XM_017020670.1
XP_016876159.1
8
1626
XM_017020671.1
XP_016876160.1
8
1549
XM_017020672.1
XP_016876161.1
8
1549
GenBank mRNA
AF183426.1
-
14
1157
AK000888.1
-
12
1112
AK001185.1
-
14
1172
AK001302.1
-
14
1141
AK002167.1
-
14
1325
AK023313.1
-
14
1161
AK024478.1
-
7
3701
AK098706.1
-
14
1175
AK289462.1
-
14
1168
BC016614.1
-
14
1160
3869275
BC031246.1
-
14
1719
5266649
BI763234.1
-
5
440
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of apoptotic process
regulation of mRNA stability
positive regulation of B cell differentiation
positive regulation of transcription, DNA-templated
spleen development
positive regulation of mitotic cell cycle spindle assembly checkpoint
negative regulation of cysteine-type endopeptidase activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0