Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ALG13 SIRNA Assays ›
Gene Symbol:
ALG13
Gene Name:
ALG13, UDP-N-acetylglucosaminyltransferase subunit
Gene Aliases:
CDG1S, CXorf45, EIEE36, GLT28D1, MDS031, TDRD13, YGL047W
Chromosome Location:
Chr.X: 111680741 - 111764164 on Build GRCh38
UniGene ID:
Hs.443061
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS125166
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
459.00
Your Price
Online offer:
459.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ALG13
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001039210.4
NP_001034299.3
3
329
NM_001099922.2
NP_001093392.1
3
388
NM_001168385.2
NP_001161857.1
3
388
NM_001257230.1
NP_001244159.1
3
284
NM_001257231.1
NP_001244160.1
3
402
NM_001257234.1
NP_001244163.1
3
357
NM_001257235.2
NP_001244164.1
3
357
NM_001257237.1
NP_001244166.1
3
461
NM_001257239.2
NP_001244168.1
3
376
NM_001257240.2
NP_001244169.1
3
461
NM_001257241.2
NP_001244170.1
3
402
NM_001324290.1
NP_001311219.1
3
394
NM_001324291.1
NP_001311220.1
3
284
NM_001324292.1
NP_001311221.1
3
388
NM_001324293.1
NP_001311222.1
3
461
NM_001324294.1
NP_001311223.1
3
272
NM_018466.5
NP_060936.1
3
388
NR_033125.2
3
284
XM_006724693.2
XP_006724756.1
3
761
XM_006724697.3
XP_006724760.2
3
661
XM_006724698.3
XP_006724761.2
3
786
XM_011531028.2
XP_011529330.1
3
762
XM_011531030.1
XP_011529332.1
3
760
XM_011531031.1
XP_011529333.1
3
461
XM_011531032.1
XP_011529334.1
3
267
XM_011531033.2
XP_011529335.1
3
307
XM_017029846.1
XP_016885335.1
3
763
XM_017029847.1
XP_016885336.1
3
762
XM_017029848.1
XP_016885337.1
3
760
XM_017029849.1
XP_016885338.1
3
761
XM_017029850.1
XP_016885339.1
3
760
XM_017029851.1
XP_016885340.1
3
763
GenBank mRNA
AF220051.1
-
3
319
AK300394.1
-
3
226
AK302729.1
-
3
329
AK302890.1
-
3
305
AK304712.1
-
3
278
AK311608.1
-
3
268
AK312229.1
-
3
327
AK316306.1
-
3
330
AK316522.1
-
3
405
AL524204.3
-
3
207
BC005336.1
-
3
321
3996922
BE747995.1
-
3
317
BF239206.1
-
3
356
BF240663.1
-
3
174
BF699152.1
-
3
390
BG621042.1
-
2
171
BM759764.1
-
3
370
BQ007420.1
-
2
346
CB268871.1
-
3
335
DA830058.1
-
3
452
DB292609.1
-
3
461
DC400909.1
-
3
272
DN992019.1
-
3
456
Back To Top
More Information
Panther Classification:
Molecular Function -
cysteine protease
transferase
Gene Ontology Categories:
Function(s)
Process(es)
dolichol-linked oligosaccharide biosynthetic process
proteolysis
N-acetylglucosaminyldiphosphodolichol N-acetylglucosaminyltransferase activity
thiol-dependent ubiquitinyl hydrolase activity
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-sskjb:80/100.66.75.127:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0