Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANKS1B SIRNA Assays ›
Gene Symbol:
ANKS1B
Gene Name:
ankyrin repeat and sterile alpha motif domain containing 1B
Gene Aliases:
AIDA, AIDA-1, ANKS2, EB-1, EB1, cajalin-2
Chromosome Location:
Chr.12: 98729904 - 99984727 on Build GRCh38
UniGene ID:
Hs.506458
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS125694
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANKS1B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001204065.1
NP_001190994.1
8
868
NM_001204066.1
NP_001190995.1
9
735
NM_001204067.1
NP_001190996.1
9
829
NM_001204068.1
NP_001190997.1
9
1292
NM_001204069.1
NP_001190998.1
9
1184
NM_001204070.1
NP_001190999.1
8
1112
NM_001204079.1
NP_001191008.1
8
871
NM_001204080.1
NP_001191009.1
7
796
NM_001204081.1
NP_001191010.1
7
796
NM_020140.3
NP_064525.1
8
1112
NM_152788.4
NP_690001.3
23
3724
NM_181670.3
NP_858056.2
10
1364
XM_005269028.4
XP_005269085.1
23
3335
XM_005269029.4
XP_005269086.1
23
3295
XM_005269032.3
XP_005269089.1
7
660
XM_006719504.3
XP_006719567.1
24
3869
XM_006719505.3
XP_006719568.1
24
3872
XM_006719506.3
XP_006719569.1
24
3869
XM_006719507.3
XP_006719570.1
24
3872
XM_006719508.3
XP_006719571.1
24
3872
XM_006719509.3
XP_006719572.1
23
3797
XM_006719510.3
XP_006719573.1
23
3797
XM_006719512.3
XP_006719575.1
22
3617
XM_006719513.3
XP_006719576.1
22
3617
XM_006719514.3
XP_006719577.1
22
3605
XM_011538571.2
XP_011536873.1
24
3770
XM_017019651.1
XP_016875140.1
24
3872
XM_017019652.1
XP_016875141.1
24
3860
XM_017019653.1
XP_016875142.1
24
3869
XM_017019654.1
XP_016875143.1
24
3857
XM_017019655.1
XP_016875144.1
24
3869
XM_017019656.1
XP_016875145.1
24
3767
XM_017019657.1
XP_016875146.1
23
3695
XM_017019658.1
XP_016875147.1
22
3617
XM_017019659.1
XP_016875148.1
10
1363
XM_017019660.1
XP_016875149.1
9
1288
XM_017019661.1
XP_016875150.1
11
1074
XM_017019662.1
XP_016875151.1
11
1207
XM_017019663.1
XP_016875152.1
10
1258
XM_017019664.1
XP_016875153.1
10
944
XM_017019665.1
XP_016875154.1
8
1108
GenBank mRNA
AF145204.1
-
17
2428
AF164792.1
-
8
1045
AK289768.1
-
10
1329
AK294191.1
-
9
829
AK294994.1
-
7
668
AK295988.1
-
8
841
AK315954.1
-
8
723
AM980945.1
-
8
727
AY281131.1
-
23
3310
AY281132.1
-
9
985
AY283057.2
-
15
2161
AY356353.1
-
7
625
AY620824.1
-
14
2035
AY753193.1
-
8
805
BC026313.1
-
10
1259
BC068451.1
-
9
735
BC142669.1
-
8
1059
BC150204.1
-
9
1131
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of synaptic plasticity by receptor localization to synapse
ephrin receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-p5ps9:80/100.66.76.38:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0