Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: APMAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020531.2 | NP_065392.1 | 3 | 341 | ||
XM_005260763.3 | XP_005260820.1 | 3 | 482 | |||
GenBank mRNA | AB033767.1 | - | 3 | 280 | ||
AK291129.1 | - | 3 | 302 | |||
AK301959.1 | - | 3 | 482 | |||
AY359076.1 | - | 2 | 128 | |||
BC003501.1 | - | 3 | 265 | 3530962 | ||
BG773501.1 | - | 3 | 538 | |||
CF131759.1 | - | 3 | 310 |