Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ADAMTSL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301110.1 | NP_001288039.1 | 26 | 4521 | ||
NM_207517.2 | NP_997400.2 | 26 | 4521 | |||
XM_011521822.2 | XP_011520124.1 | 27 | 4593 | |||
XM_011521823.2 | XP_011520125.1 | 27 | 4593 | |||
XM_011521824.2 | XP_011520126.1 | 27 | 4593 | |||
XM_017022434.1 | XP_016877923.1 | 27 | 4593 | |||
XM_017022435.1 | XP_016877924.1 | 26 | 4389 | |||
GenBank mRNA | AB033059.1 | - | 10 | 2295 | ||
AK308845.1 | - | 26 | 4512 | |||
BC128389.1 | - | 26 | 4519 | 40125494 | ||
BC128390.1 | - | 26 | 4519 | 40125501 |