Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: LRCH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243963.1 | NP_001230892.1 | 7 | 1073 | ||
NM_020871.3 | NP_065922.3 | 7 | 1073 | |||
XM_006724724.3 | XP_006724787.2 | 7 | 1076 | |||
XM_017029696.1 | XP_016885185.1 | 7 | 1082 | |||
XM_017029697.1 | XP_016885186.1 | 7 | 1083 | |||
GenBank mRNA | AB040928.1 | - | 3 | 254 | ||
BC125224.1 | - | 7 | 1066 | 40124466 | ||
BX647193.1 | - | 7 | 1073 |