Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RFWD2 SIRNA Assays ›
          Gene Symbol:
          RFWD2
          Gene Name:
          ring finger and WD repeat domain 2
          Gene Aliases:
          COP1, RNF200
          Chromosome Location:
          Chr.1: 175944826 - 176207279 on Build GRCh38
          UniGene ID:
          Hs.523744
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS127527
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RFWD2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001001740.3 NP_001001740.1 13, 14 1815
          NM_001286644.1 NP_001273573.1 12, 13 1690
          NM_022457.6 NP_071902.2 14, 15 1887
          XM_005245447.2 XP_005245504.1 14, 15 1910
          XM_005245448.2 XP_005245505.1 13, 14 1862
          XM_006711487.2 XP_006711550.1 13, 14 1853
          XM_011509881.1 XP_011508183.1 15, 16 1976
          XM_017002059.1 XP_016857548.1 15, 16 1976
          XM_017002060.1 XP_016857549.1 15, 16 1964
          XM_017002061.1 XP_016857550.1 14, 15 1922
          XM_017002062.1 XP_016857551.1 14, 15 1916
          XM_017002063.1 XP_016857552.1 14, 15 1910
          XM_017002064.1 XP_016857553.1 14, 15 1907
          XM_017002065.1 XP_016857554.1 13, 14 1862
          XM_017002066.1 XP_016857555.1 13, 14 1853
          XM_017002067.1 XP_016857556.1 13, 14 1850
          XM_017002068.1 XP_016857557.1 13, 14 1847
          XM_017002069.1 XP_016857558.1 9, 10 1490
          XM_017002070.1 XP_016857559.1 14, 15 1682
          XM_017002071.1 XP_016857560.1 12, 13 1071
          XM_017002072.1 XP_016857561.1 12, 13 1069
          XM_017002073.1 XP_016857562.1 11, 12 1011
          XM_017002074.1 XP_016857563.1 11, 12 1009
          XM_017002075.1 XP_016857564.1 11, 12 1091
          XM_017002076.1 XP_016857565.1 12, 13 1074
          XM_017002077.1 XP_016857566.1 10, 11 866
          XM_017002078.1 XP_016857567.1 12, 13 1139
          XM_017002079.1 XP_016857568.1 12, 13 1554
          XM_017002080.1 XP_016857569.1 8, 9 793
          GenBank mRNA AF508940.1 - 14, 15 1621
          AF527539.1 - 14, 15 1884
          AK001278.1 - 12, 13 1125
          AK314750.1 - 14, 15 1819
          AY509921.1 - 13, 14 1549
          BC039723.1 - 12, 13 1137
          BC094728.1 - 14, 15 1875 5271629
          BX537932.1 - 3, 4 285
          KJ534928.1 - 14, 15 1621
          KJ534929.1 - 11, 12 1426
          KJ534930.1 - 13, 14 1561
          KJ534931.1 - 13, 14 1552
          KJ534932.1 - 14, 15 1609
          KJ534933.1 - 8, 9 1120
          KJ535061.1 - 11, 12 1326
          KJ535076.1 - 13, 14 1523

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to ionizing radiation
          protein ubiquitination
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          ubiquitin-protein transferase activity
          protein binding
          zinc ion binding
          ligase activity
          ubiquitin protein ligase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0