Search
Search

NFKBIZ| Gene Symbol: NFKBIZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005474.2 | NP_001005474.1 | 9 | 1594 | ||
| NM_031419.3 | NP_113607.1 | 8 | 1727 | |||
| GenBank mRNA | AB037925.1 | - | 8 | 1660 | ||
| AF548362.1 | - | 9 | 1591 | |||
| AK054787.1 | - | 9 | 1463 | |||
| AK091782.1 | - | 9 | 1361 | |||
| BC014587.2 | - | 1373 | ||||
| BC060800.1 | - | 8 | 1727 | 30344780 | ||
| DQ224339.1 | - | 8 | 1516 | |||