Search Thermo Fisher Scientific
Gene Symbol: CYP3A43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278921.1 | NP_001265850.1 | 5 | 487 | ||
NM_022820.4 | NP_073731.1 | 8 | 817 | |||
NM_057095.2 | NP_476436.1 | 8 | 817 | |||
NM_057096.3 | NP_476437.1 | 8 | 817 | |||
NR_103869.1 | 9 | 938 | ||||
XM_017012542.1 | XP_016868031.1 | 6 | 485 | |||
XM_017012543.1 | XP_016868032.1 | 5 | 643 | |||
XM_017012544.1 | XP_016868033.1 | 4 | 371 | |||
XM_017012545.1 | XP_016868034.1 | 4 | 351 | |||
GenBank mRNA | AF280108.1 | - | 8 | 714 | ||
AF280109.1 | - | 8 | 714 | |||
AF280110.1 | - | 8 | 714 | |||
AF280111.1 | - | 9 | 835 | |||
AF280112.1 | - | 5 | 394 | |||
AF319634.1 | - | 8 | 714 | |||
AF337813.1 | - | 8 | 817 | |||
AY390423.1 | - | 8 | 713 | |||
AY390424.1 | - | 8 | 714 | |||
AY390425.1 | - | 8 | 714 | |||
AY390426.1 | - | 8 | 713 | |||
BC100981.3 | - | 5 | 457 | 40005401 |