Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ELOVL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130721.1 | NP_001124193.1 | 4 | 471 | ||
NM_024090.2 | NP_076995.1 | 3 | 674 | |||
XM_011532233.2 | XP_011530535.1 | 4 | 477 | |||
XM_011532234.2 | XP_011530536.1 | 4 | 545 | |||
XM_011532235.2 | XP_011530537.1 | 4 | 589 | |||
GenBank mRNA | AK027031.1 | - | 3 | 625 | ||
AK074813.1 | - | 3 | 319 | |||
BC001305.1 | - | 4 | 458 | 3452423 | ||
BP314858.1 | - | 4 | 471 |