Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: PLEKHF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024310.4 | NP_077286.3 | 2 | 120 | ||
XM_005259256.3 | XP_005259313.1 | 2 | 408 | |||
XM_011527309.2 | XP_011525611.1 | 2 | 1394 | |||
GenBank mRNA | AF434818.1 | - | 1 | 18 | ||
AK027758.1 | - | 2 | 79 | |||
AK225814.1 | - | 2 | 352 | |||
AK301616.1 | - | 2 | 372 | |||
AY037145.2 | - | 2 | 484 | |||
BC002744.2 | - | 2 | 74 | 3628796 | ||
BC018876.2 | - | 2 | 74 |