Search
Search

CHPF| Gene Symbol: CHPF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195731.1 | NP_001182660.1 | 4 | 1788 | ||
| NM_024536.5 | NP_078812.2 | 4 | 2144 | |||
| XM_011511838.2 | XP_011510140.1 | 3 | 1675 | |||
| GenBank mRNA | AB086063.1 | - | 4 | 1978 | ||
| AB095813.1 | - | 4 | 2153 | |||
| AK026331.1 | - | 1 | 568 | |||
| AK302124.1 | - | 4 | 1788 | |||
| AL136814.1 | - | 3 | 1291 | |||
| AY358403.1 | - | 4 | 2144 | |||
| BC008878.2 | - | 1 | 696 | |||
| BC021223.2 | - | 4 | 2144 | 4128192 | ||
| BC023531.2 | - | 4 | 2095 | 4123041 | ||