Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NEIL1 SIRNA Assays ›
          Gene Symbol:
          NEIL1
          Gene Name:
          nei like DNA glycosylase 1
          Gene Aliases:
          FPG1, NEI1, hFPG1
          Chromosome Location:
          Chr.15: 75346895 - 75355251 on Build GRCh38
          UniGene ID:
          Hs.512732
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS128612
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NEIL1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256552.1 NP_001243481.1 2 513
          NM_024608.3 NP_078884.2 2 589
          NR_046311.1 3 712
          XM_005254659.4 XP_005254716.1 2 449
          XM_006720677.3 XP_006720740.1 2 434
          XM_006720678.3 XP_006720741.1 2 464
          XM_006720679.3 XP_006720742.1 2 372
          XM_006720680.1 XP_006720743.1 3 604
          XM_006720681.1 XP_006720744.1 3 451
          XM_011522001.2 XP_011520303.2 2 354
          XM_011522002.1 XP_011520304.1 3 577
          XM_011522003.2 XP_011520305.1 4 633
          XM_011522004.2 XP_011520306.1 2 321
          XM_011522005.1 XP_011520307.1 2 508
          XM_011522006.1 XP_011520308.1 2 508
          XM_011522007.1 XP_011520309.1 3 604
          XM_011522008.1 XP_011520310.1 3 577
          XM_017022566.1 XP_016878055.1 2 436
          XM_017022567.1 XP_016878056.1 2 268
          XM_017022568.1 XP_016878057.1 2 471
          GenBank mRNA AB079068.1 - 1 83
          AK026055.1 - 2 510
          AK097008.1 - 3 712
          AK128372.1 - 2 228
          BC010876.1 - 2 494 3879198

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA glycosylase

          Gene Ontology Categories:

          Function(s) Process(es)

          base-excision repair
          nucleotide-excision repair
          response to oxidative stress
          negative regulation of nuclease activity
          depyrimidination
          damaged DNA binding
          DNA-(apurinic or apyrimidinic site) lyase activity
          protein C-terminus binding
          zinc ion binding
          hydrolase activity, acting on glycosyl bonds
          DNA N-glycosylase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0