Search
Search

FN3KRP| Gene Symbol: FN3KRP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024619.3 | NP_078895.2 | 6 | 792 | ||
| NR_046408.1 | 7 | 920 | ||||
| GenBank mRNA | AK022233.1 | - | 6 | 759 | ||
| AK293523.1 | - | 5 | 695 | |||
| AK315373.1 | - | 6 | 776 | |||
| AL136631.1 | - | 6 | 763 | |||
| AY360465.1 | - | 6 | 759 | |||
| BC001458.2 | - | 3 | 330 | |||
| BC007611.1 | - | 6 | 768 | 3351601 | ||
| BC014408.1 | - | 6 | 746 | 4652087 | ||