Search
Search

GALNT12| Gene Symbol: GALNT12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024642.4 | NP_078918.3 | 10 | 1675 | ||
| XM_006717287.1 | XP_006717350.1 | 10 | 1491 | |||
| XM_011519018.2 | XP_011517320.1 | 11 | 2347 | |||
| XM_011519020.1 | XP_011517322.1 | 10 | 1523 | |||
| XM_017015133.1 | XP_016870622.1 | 11 | 1435 | |||
| GenBank mRNA | AB040673.1 | - | 10 | 1675 | ||
| AB078146.1 | - | 10 | 1675 | |||
| AJ132365.2 | - | 10 | 1675 | |||
| AJ505963.1 | - | 4 | 454 | |||
| AK024865.1 | - | 9 | 1248 | |||
| BC013945.1 | - | 7 | 840 | |||