Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TBC1D17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024682.2 | NP_078958.2 | 2 | 332 | ||
XM_011527317.2 | XP_011525619.1 | 2 | 244 | |||
GenBank mRNA | AB449903.1 | - | 2 | 33 | ||
AK022230.1 | - | 2 | 91 | |||
AK090606.1 | - | 1 | 19 | |||
AK309412.1 | - | 2 | 57 | |||
BC003516.1 | - | 2 | 185 | 3050014 |