Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: C10orf88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024942.3 | NP_079218.2 | 4 | 845 | ||
GenBank mRNA | AK023552.1 | - | 4 | 701 | ||
BC032645.1 | - | 4 | 685 | 5519534 |