Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CCDC130 SIRNA Assays ›
          Gene Symbol:
          CCDC130
          Gene Name:
          coiled-coil domain containing 130
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 13731760 - 13763298 on Build GRCh38
          UniGene ID:
          Hs.24998
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS129944
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CCDC130
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001320561.1 NP_001307490.1 4, 5 696
          NM_001320564.1 NP_001307493.1 4, 5 684
          NM_001320565.1 NP_001307494.1 4, 5 566
          NM_001320566.1 NP_001307495.1 4, 5 684
          NM_001320567.1 NP_001307496.1 4, 5 547
          NM_001320568.1 NP_001307497.1 3, 4 356
          NM_001320569.1 NP_001307498.1 2, 3 152
          NM_030818.3 NP_110445.1 3, 4 356
          XM_005260086.4 XP_005260143.1 4, 5 546
          XM_011528326.1 XP_011526628.1 3, 4 435
          GenBank mRNA AF250306.1 - 3, 4 284
          AK098262.1 - 3, 4 313
          AK293927.1 - 4, 5 547
          AL560878.3 - 4, 5 566
          BC002905.2 - 3, 4 280 3940130
          BI913255.1 - 2, 3 117
          BM547551.1 - 3, 4 354
          BX394050.2 - 4, 5 677
          BX422326.2 - 3, 4 355
          CR999732.1 - 2, 3 281

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          RNA splicing factor

          Gene Ontology Categories:

          Function(s) Process(es)

          response to virus
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-jww4n:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0