Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FBXO18 SIRNA Assays ›
          Gene Symbol:
          FBXO18
          Gene Name:
          F-box protein, helicase, 18
          Gene Aliases:
          FBH1, Fbx18, hFBH1
          Chromosome Location:
          Chr.10: 5889572 - 5937595 on Build GRCh38
          UniGene ID:
          Hs.498543
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS131402
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FBXO18
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001258452.1 NP_001245381.1 17 2625
          NM_001258453.1 NP_001245382.1 18 2697
          NM_032807.4 NP_116196.3 18 2713
          NM_178150.2 NP_835363.1 17 2599
          XM_011519748.2 XP_011518050.1 19 3293
          XM_017016842.1 XP_016872331.1 18 3840
          XM_017016843.1 XP_016872332.1 19 3224
          XM_017016844.1 XP_016872333.1 17 2609
          XM_017016845.1 XP_016872334.1 17 2966
          XM_017016846.1 XP_016872335.1 16 2418
          GenBank mRNA AF380349.1 - 18 2713
          AF454502.1 - 17 2540
          AF456237.1 - 17 2555
          AK027381.1 - 8 719
          AK027496.1 - 14 1682
          AK095343.1 - 17 2573
          AK122753.1 - 17 2625
          AK226128.1 - 17 3123
          AK315173.1 - 16 2315
          AL133069.1 - 3 172
          BC012762.2 - 4 341
          BC020266.1 - 15 2107
          BC032674.1 - 15 2051
          BC080629.1 - 15 2262
          BC110884.1 - 18 2622 6147373
          BC113375.1 - 17 2546 8322427
          BC113377.1 - 17 2546 8322429

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          double-strand break repair via homologous recombination
          DNA catabolic process, endonucleolytic
          positive regulation of protein phosphorylation
          cellular response to DNA damage stimulus
          cell death
          protein ubiquitination
          replication fork processing
          DNA duplex unwinding
          negative regulation of chromatin binding
          replication fork protection
          response to intra-S DNA damage checkpoint signaling
          positive regulation of intrinsic apoptotic signaling pathway in response to DNA damage
          negative regulation of double-strand break repair via homologous recombination
          DNA helicase activity
          double-stranded DNA binding
          single-stranded DNA binding
          ATP-dependent DNA helicase activity
          protein binding
          ATP binding
          DNA translocase activity
          3'-5' DNA helicase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0