Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF566 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145343.1 | NP_001138815.1 | 4, 5 | 375 | ||
NM_001145344.1 | NP_001138816.1 | 4, 5 | 372 | |||
NM_001145345.1 | NP_001138817.1 | 4, 5 | 384 | |||
NM_001300970.1 | NP_001287899.1 | 3, 4 | 598 | |||
NM_032838.4 | NP_116227.1 | 4, 5 | 319 | |||
XM_006723447.2 | XP_006723510.1 | 4, 5 | 728 | |||
XM_017027399.1 | XP_016882888.1 | 3, 4 | 586 | |||
GenBank mRNA | AK027685.1 | - | 4, 5 | 319 | ||
AK074497.1 | - | 4, 5 | 329 | |||
BC007064.2 | - | 3, 4 | 296 | 3961390 | ||
BC104886.1 | - | 3, 4 | 291 | 8143889 | ||
BC104888.1 | - | 3, 4 | 291 | 8143891 | ||
BC143658.1 | - | 3, 4 | 294 | 9052172 | ||
BP351422.1 | - | 4, 5 | 384 | |||
DC373988.1 | - | 4, 5 | 350 | |||
DC413210.1 | - | 4, 5 | 376 | |||
HY178567.1 | - | 3, 4 | 233 |