Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: C11orf70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195005.1 | NP_001181934.1 | 4 | 440 | ||
NM_032930.2 | NP_116319.2 | 5 | 607 | |||
XM_005271713.3 | XP_005271770.1 | 5 | 635 | |||
GenBank mRNA | AK094851.1 | - | 5 | 587 | ||
BC006128.1 | - | 4 | 811 | 3622924 | ||
BC054869.1 | - | 4 | 426 |