Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: EPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB051511.1 | - | 9 | 1035 | ||
AK095620.1 | - | 9 | 1045 | |||
BC021229.2 | - | 9 | 898 | 4111329 |