Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MLIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281746.1 | NP_001268675.1 | 2 | 208 | ||
NM_001281747.1 | NP_001268676.1 | 2 | 190 | |||
NM_138569.2 | NP_612636.2 | 2 | 208 | |||
XM_005249476.4 | XP_005249533.1 | 2 | 161 | |||
XM_005249480.4 | XP_005249537.1 | 2 | 161 | |||
XM_006715245.3 | XP_006715308.2 | 2 | 300 | |||
XM_017011465.1 | XP_016866954.1 | 2 | 297 | |||
XM_017011466.1 | XP_016866955.1 | 2 | 294 | |||
XM_017011467.1 | XP_016866956.1 | 2 | 303 | |||
XM_017011468.1 | XP_016866957.1 | 2 | 302 | |||
XM_017011469.1 | XP_016866958.1 | 2 | 94 | |||
XM_017011470.1 | XP_016866959.1 | 2 | 392 | |||
GenBank mRNA | AK055530.1 | - | 2 | 133 | ||
AK294890.1 | - | 2 | 161 | |||
AK311190.1 | - | 2 | 161 | |||
AK316157.1 | - | 2 | 141 | |||
BX091237.1 | - | 2 | 75 | |||
BX647567.1 | - | 2 | 154 | |||
DA761851.1 | - | 2 | 208 | |||
KU178823.1 | - | 2 | 65 | |||
KU178824.1 | - | 2 | 65 | |||
KU178825.1 | - | 2 | 65 | |||
KU178826.1 | - | 2 | 65 |