Search
Search

FCRL1| Gene Symbol: FCRL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159398.1 | NP_001152870.1 | 6 | 1019 | ||
| NM_052938.4 | NP_443170.1 | 6 | 1019 | |||
| XM_005244866.2 | XP_005244923.1 | 6 | 990 | |||
| XM_005244869.3 | XP_005244926.1 | 4 | 715 | |||
| XM_011509125.1 | XP_011507427.1 | 6 | 990 | |||
| XM_011509126.1 | XP_011507428.1 | 6 | 990 | |||
| XM_011509127.2 | XP_011507429.1 | 5 | 982 | |||
| XM_011509128.1 | XP_011507430.1 | 6 | 990 | |||
| XM_011509129.1 | XP_011507431.1 | 6 | 990 | |||
| XM_011509130.1 | XP_011507432.1 | 6 | 990 | |||
| XM_011509131.1 | XP_011507433.1 | 6 | 990 | |||
| XM_011509132.2 | XP_011507434.1 | 6 | 990 | |||
| XM_011509133.2 | XP_011507435.1 | 6 | 844 | |||
| XM_011509134.2 | XP_011507436.1 | 5 | 819 | |||
| XM_011509135.2 | XP_011507437.1 | 5 | 736 | |||
| XM_011509136.1 | XP_011507438.1 | 6 | 1004 | |||
| XM_011509137.2 | XP_011507439.1 | 3 | 364 | |||
| XM_017000227.1 | XP_016855716.1 | 5 | 2986 | |||
| GenBank mRNA | AF329488.1 | - | 6 | 929 | ||
| AF459634.1 | - | 6 | 972 | |||
| AK315748.1 | - | 6 | 947 | |||
| AL833674.1 | - | 2 | 73 | |||
| AY043464.1 | - | 6 | 974 | |||
| BC021274.2 | - | 6 | 687 | |||
| BC033690.1 | - | 6 | 970 | 5211491 | ||