Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TM4SF19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204897.1 | NP_001191826.1 | 2 | 278 | ||
NM_001204898.1 | NP_001191827.1 | 2 | 278 | |||
NM_138461.3 | NP_612470.2 | 2 | 278 | |||
GenBank mRNA | AF192910.1 | - | 2 | 234 | ||
AK310209.1 | - | 2 | 248 | |||
BC146985.1 | - | 2 | 179 | 9056913 | ||
BC146993.1 | - | 2 | 179 | 9056921 | ||
BC147021.1 | - | 2 | 179 | 9056949 | ||
BC171870.1 | - | 2 | 179 | 9054524 | ||
BQ678650.1 | - | 2 | 244 | |||
CA489370.1 | - | 2 | 278 | |||
DB015357.1 | - | 2 | 248 |
Gene Symbol: TM4SF19-TCTEX1D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037950.1 | 2 | 278 |